Gene Transcripts
- Gene Symbol
- WDR97
- Entrez Gene ID
- 340390
- Gene Name
- WD repeat domain 97
- Gene Aliases
- KIAA1875
- Chromosome Location
- Chr. 8: 144107692 - 144116392 on Build GRCh38
- UniGene ID
- Hs.98723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316309.1 |
NP_001303238.1
|
9, 10 |
2355 |
|
XM_017013385.1 |
XP_016868874.1
|
9, 10 |
2349 |
|
XM_017013386.1 |
XP_016868875.1
|
9, 10 |
2349 |
|
XM_017013387.1 |
XP_016868876.1
|
9, 10 |
2290 |
|
XM_017013388.1 |
XP_016868877.1
|
9, 10 |
2349 |
|
XM_017013389.1 |
XP_016868878.1
|
9, 10 |
2349 |
|
XM_017013390.1 |
XP_016868879.1
|
9, 10 |
2349 |
|
XM_017013391.1 |
XP_016868880.1
|
9, 10 |
2349 |
|
XM_017013392.1 |
XP_016868881.1
|
9, 10 |
2349 |
|
XM_017013393.1 |
XP_016868882.1
|
9, 10 |
2349 |
|
XM_017013394.1 |
XP_016868883.1
|
8, 9 |
2135 |
|
GenBank mRNA |
AB058778.1 |
- |
9, 10 |
2321 |
|
AK095597.1 |
- |
3, 4 |
1087 |
|
AK302485.1 |
- |
5, 6 |
876 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB097314.1 |
- |
|
576 |
|
Chr. 8 - 144110106 - 144110996 [+] |
GenBankmRNA |
DB064296.1 |
- |
1, 2 |
88 |
|
Chr. 8 - 144110901 - 144111525 [+] |
View Full Product Details