Gene Transcripts
- Gene Symbol
- LINC00626
- Entrez Gene ID
- 79100
- Gene Name
- long intergenic non-protein coding RNA 626
- Gene Aliases
- Chromosome Location
- Chr. 1: 168786941 - 168792888 on Build GRCh38
- UniGene ID
- Hs.250624
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024160.1 |
|
3 |
723 |
|
GenBank mRNA |
BC068047.1 |
- |
3 |
590 |
|
BC082964.1 |
- |
3 |
590 |
|
View Full Product Details