Gene Transcripts

Gene Symbol
SNHG6
Entrez Gene ID
641638
Gene Name
small nucleolar RNA host gene 6
Gene Aliases
HBII-276HG, NCRNA00058, U87HG
Chromosome Location
Chr. 8: 66921930 - 66925542 on Build GRCh38
UniGene ID
Hs.372680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002599.1 2 151
GenBank mRNA AY264285.1 - 2 151


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI669107.1 - 2 206 Chr. 8 - 66921934 - 66925598 [-]
GenBankmRNA HY162808.1 - 2 150 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA BF813358.1 - 2 87 Chr. 8 - 66921949 - 66925459 [-]
GenBankmRNA BM737762.1 - 2 138 Chr. 8 - 66921984 - 66925529 [-]
GenBankmRNA BQ924387.1 - 128 Chr. - - [-]
GenBankmRNA AA522925.1 - 2 142 Chr. 8 - 66921986 - 66925531 [-]
GenBankmRNA BI769050.1 - 133 Chr. - - [-]
GenBankmRNA EL946830.1 - 2 118 Chr. 8 - 66922045 - 66925495 [-]
GenBankmRNA BI669733.1 - 2 155 Chr. 8 - 66921925 - 66925541 [-]
GenBankmRNA DA126867.1 - 2 150 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA BM016905.1 - 2 135 Chr. 8 - 66921935 - 66925526 [-]
GenBankmRNA BY796444.2 - 2 126 Chr. 8 - 66921935 - 66925517 [-]
GenBankmRNA BM737568.1 - 149 Chr. - - [-]
GenBankmRNA CB130641.1 - 2 150 Chr. 8 - 66922002 - 66925541 [-]
GenBankmRNA F20193.2 - 123 Chr. - - [-]
GenBankmRNA BU193261.1 - 2 148 Chr. 8 - 66921934 - 66925530 [-]
GenBankmRNA BU167862.1 - 2 151 Chr. 8 - 66921925 - 66925534 [-]
GenBankmRNA BU193032.1 - 2 117 Chr. 8 - 66921934 - 66925508 [-]
GenBankmRNA BU195948.1 - 2 133 Chr. 8 - 66921968 - 66925524 [-]
GenBankmRNA CN257859.1 - 2 128 Chr. 8 - 66921938 - 66925519 [-]
GenBankmRNA BQ931992.1 - 154 Chr. - - [-]
GenBankmRNA AJ712674.1 - 2 125 Chr. 8 - 66922010 - 66925516 [-]
GenBankmRNA BQ267112.1 - 2 122 Chr. 8 - 66921995 - 66925506 [-]
GenBankmRNA AA633432.1 - 2 135 Chr. 8 - 66921984 - 66925524 [-]
GenBankmRNA BG034409.1 - 147 Chr. - - [-]
GenBankmRNA HY072040.1 - 2 149 Chr. 8 - 66921934 - 66925540 [-]
GenBankmRNA BM014460.1 - 2 134 Chr. 8 - 66921934 - 66925525 [-]
GenBankmRNA AA307876.1 - 2 134 Chr. 8 - 66922048 - 66925525 [-]
GenBankmRNA BM784351.1 - 2 150 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA W56516.1 - 118 Chr. - - [-]
GenBankmRNA BF966290.2 - 2 155 Chr. 8 - 66921934 - 66925541 [-]
GenBankmRNA DB498305.1 - 2 201 Chr. 8 - 66921934 - 66925591 [-]
GenBankmRNA BM768256.1 - 2 150 Chr. 8 - 66921968 - 66925541 [-]
GenBankmRNA AW958356.1 - 135 Chr. - - [-]
GenBankmRNA BF814456.1 - 2 87 Chr. 8 - 66921949 - 66925459 [-]
GenBankmRNA AA429085.1 - 2 139 Chr. 8 - 66921958 - 66925527 [-]
GenBankmRNA BU596037.1 - 153 Chr. - - [-]
GenBankmRNA BQ440073.1 - 132 Chr. - - [-]
GenBankmRNA BF212415.1 - 2 155 Chr. 8 - 66921933 - 66925545 [-]
GenBankmRNA BM743937.1 - 2 149 Chr. 8 - 66921990 - 66925541 [-]
GenBankmRNA BI918140.1 - 136 Chr. - - [-]
GenBankmRNA CB963297.1 - 2 95 Chr. 8 - 66921984 - 66925481 [-]
GenBankmRNA HY294280.1 - 2 190 Chr. 8 - 66922000 - 66925542 [-]
GenBankmRNA BG032838.1 - 120 Chr. - - [-]
GenBankmRNA DB471689.1 - 2 132 Chr. 8 - 66921984 - 66925523 [-]
GenBankmRNA HY117390.1 - 2 201 Chr. 8 - 66921984 - 66925591 [-]
GenBankmRNA BG034121.1 - 2 140 Chr. 8 - 66921938 - 66925531 [-]
GenBankmRNA BM740598.1 - 2 150 Chr. 8 - 66922322 - 66925541 [-]
GenBankmRNA BM842017.1 - 2 151 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA BM842145.1 - 2 151 Chr. 8 - 66921984 - 66925542 [-]
GenBankmRNA HY279249.1 - 2 187 Chr. 8 - 66922022 - 66925541 [-]
GenBankmRNA BE254660.1 - 114 Chr. - - [-]
GenBankmRNA BI550715.1 - 2 156 Chr. 8 - 66921934 - 66925542 [-]
GenBankmRNA BQ073523.1 - 128 Chr. - - [-]
GenBankmRNA BP425366.1 - 2 127 Chr. 8 - 66921938 - 66925511 [-]
GenBankmRNA HY066133.1 - 2 149 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA BM741967.1 - 2 149 Chr. 8 - 66921991 - 66925541 [-]
GenBankmRNA BU565593.1 - 2 153 Chr. 8 - 66921984 - 66925541 [-]
GenBankmRNA AW630767.1 - 2 127 Chr. 8 - 66921971 - 66925518 [-]
GenBankmRNA HY112530.1 - 2 150 Chr. 8 - 66921948 - 66925540 [-]
GenBankmRNA HY075045.1 - 2 152 Chr. 8 - 66921984 - 66925540 [-]
GenBankmRNA DB569402.1 - 2 148 Chr. 8 - 66921936 - 66925539 [-]
GenBankmRNA CX752858.1 - 2 133 Chr. 8 - 66921934 - 66925524 [-]
GenBankmRNA CF129421.1 - 1 56 Chr. 8 - 66921934 - 66922723 [-]
GenBankmRNA DA301664.1 - 2 202 Chr. 8 - 66921910 - 66925593 [-]
GenBankmRNA BM016312.1 - 119 Chr. - - [-]
GenBankmRNA DA154192.1 - 2 150 Chr. 8 - 66921818 - 66925541 [-]
GenBankmRNA HY084735.1 - 2 149 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA BG703295.1 - 2 137 Chr. 8 - 66921984 - 66925525 [-]
GenBankmRNA CB122847.1 - 2 122 Chr. 8 - 66921984 - 66925513 [-]
GenBankmRNA CD513777.1 - 2 162 Chr. 8 - 66921989 - 66925530 [-]
GenBankmRNA F24069.1 - 116 Chr. 11 - 76191801 - 76191834 [-]
GenBankmRNA BU566037.1 - 107 Chr. - - [-]
GenBankmRNA BE789325.1 - 2 104 Chr. 8 - 66921991 - 66925496 [-]
GenBankmRNA BM737847.1 - 2 139 Chr. 8 - 66921984 - 66925530 [-]
GenBankmRNA CD513303.1 - 2 128 Chr. 8 - 66921984 - 66925519 [-]
GenBankmRNA BM015094.1 - 135 Chr. - - [-]
GenBankmRNA BM840736.1 - 2 151 Chr. 8 - 66921996 - 66925542 [-]
GenBankmRNA F21431.1 - 1, 2 122 Chr. 19 - 16889188 - 16890995 [+]
GenBankmRNA BE875021.1 - 2 117 Chr. 8 - 66921986 - 66925508 [-]
GenBankmRNA DB540433.1 - 2 132 Chr. 8 - 66921997 - 66925523 [-]
GenBankmRNA HY056578.1 - 2 149 Chr. 8 - 66921934 - 66925540 [-]
GenBankmRNA BM760648.1 - 2 151 Chr. 8 - 66921991 - 66925542 [-]
GenBankmRNA DA084810.1 - 2 151 Chr. 8 - 66921839 - 66925542 [-]
GenBankmRNA EL946282.1 - 2 142 Chr. 8 - 66921971 - 66925487 [-]
GenBankmRNA BM737712.1 - 2 136 Chr. 8 - 66921986 - 66925527 [-]
GenBankmRNA BU953849.1 - 2 176 Chr. 8 - 66921987 - 66925566 [-]
GenBankmRNA HY012009.1 - 2 149 Chr. 8 - 66921987 - 66925540 [-]
GenBankmRNA AA316590.1 - 2 122 Chr. 8 - 66921930 - 66925513 [-]
GenBankmRNA HY122223.1 - 2 149 Chr. 8 - 66921936 - 66925540 [-]
GenBankmRNA CB143382.1 - 2 246 Chr. 8 - 66922018 - 66926398 [-]
GenBankmRNA BQ229214.1 - 100 Chr. - - [-]
GenBankmRNA BM511306.1 - 2 106 Chr. 8 - 66921984 - 66925490 [-]

View Full Product Details