Gene Transcripts
- Gene Symbol
- SH3TC1
- Entrez Gene ID
- 54436
- Gene Name
- SH3 domain and tetratricopeptide repeats 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 8182072 - 8241103 on Build GRCh38
- UniGene ID
- Hs.479116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057561.1 |
- |
1 |
672 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB100951.1 |
- |
1 |
672 |
|
Chr. 4 - 8208564 - 8209322 [+] |
View Full Product Details