Gene Transcripts
- Gene Symbol
- LOC100506985
- Entrez Gene ID
- 100506985
- Gene Name
- uncharacterized LOC100506985
- Gene Aliases
- Chromosome Location
- Chr. 1: 24539274 - 24556024 on Build GRCh38
- UniGene ID
- Hs.607088
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109781.1 |
|
2, 3 |
374 |
|
View Full Product Details