Gene Transcripts
- Gene Symbol
- HEATR6
- Entrez Gene ID
- 63897
- Gene Name
- HEAT repeat containing 6
- Gene Aliases
- ABC1
- Chromosome Location
- Chr. 17: 60041363 - 60078931 on Build GRCh38
- UniGene ID
- Hs.221899
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BP394917.1 |
- |
1 |
186 |
|
Chr. 17 - 60041458 - 60041886 [-] |
GenBankmRNA |
BM692420.1 |
- |
1 |
150 |
|
Chr. 17 - 60041318 - 60041850 [-] |
GenBankmRNA |
CN412281.1 |
- |
1 |
181 |
|
Chr. 17 - 60041367 - 60041874 [-] |
View Full Product Details