Gene Transcripts
- Gene Symbol
- KBTBD3
- Entrez Gene ID
- 143879
- Gene Name
- kelch repeat and BTB domain containing 3
- Gene Aliases
- BKLHD3
- Chromosome Location
- Chr. 11: 106046801 - 106077765 on Build GRCh38
- UniGene ID
- Hs.101949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124540.1 |
- |
1 |
1662 |
|
View Full Product Details