Gene Transcripts
- Gene Symbol
- NCAPH
- Entrez Gene ID
- 23397
- Gene Name
- non-SMC condensin I complex subunit H
- Gene Aliases
- BRRN1, CAP-H
- Chromosome Location
- Chr. 2: 96335741 - 96377095 on Build GRCh38
- UniGene ID
- Hs.308045
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR608905.1 |
- |
1 |
1792 |
|
Chr. 2 - 96374664 - 96377067 [+] |
GenBankmRNA |
BX096707.1 |
- |
1 |
306 |
|
Chr. 2 - 96376159 - 96376810 [+] |
GenBankmRNA |
CB123155.1 |
- |
1 |
97 |
|
Chr. 2 - 96376359 - 96376660 [+] |
View Full Product Details