Gene Transcripts
- Gene Symbol
- LOC645485
- Entrez Gene ID
- 645485
- Gene Name
- uncharacterized LOC645485
- Gene Aliases
- Chromosome Location
- Chr. 12: 30755074 - 30780739 on Build GRCh38
- UniGene ID
- Hs.232332
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135046.1 |
|
2 |
263 |
|
GenBank mRNA |
BC039526.1 |
- |
2 |
263 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM563950.1 |
- |
2 |
283 |
|
Chr. 12 - 30755074 - 30780230 [+] |
View Full Product Details