Gene Transcripts
- Gene Symbol
- LOC101928992
- Entrez Gene ID
- 101928992
- Gene Name
- uncharacterized LOC101928992
- Gene Aliases
- Chromosome Location
- Chr. 3: 184756389 - 184773155 on Build GRCh38
- UniGene ID
- Hs.507129
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110043.1 |
|
3 |
1966 |
|
GenBank mRNA |
BC050282.1 |
- |
3 |
1966 |
|
View Full Product Details