Gene Transcripts
- Gene Symbol
- LOC100287042
- Entrez Gene ID
- 100287042
- Gene Name
- uncharacterized LOC100287042
- Gene Aliases
- Chromosome Location
- Chr. 17: 75271299 - 75273895 on Build GRCh38
- UniGene ID
- Hs.514470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036520.1 |
|
1 |
1683 |
|
GenBank mRNA |
BC021162.2 |
- |
1 |
362 |
|
View Full Product Details