Gene Transcripts
- Gene Symbol
- LOC100287042
- Entrez Gene ID
- 100287042
- Gene Name
- uncharacterized LOC100287042
- Gene Aliases
- Chromosome Location
- Chr. 17: 75271299 - 75273895 on Build GRCh38
- UniGene ID
- Hs.514470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036520.1 |
|
1 |
1818 |
|
GenBank mRNA |
BC021162.2 |
- |
1 |
497 |
|
- Gene Symbol
- SLC25A19
- Entrez Gene ID
- 60386
- Gene Name
- solute carrier family 25 member 19
- Gene Aliases
- DNC, MCPHA, MUP1, THMD3, THMD4, TPC
- Chromosome Location
- Chr. 17: 75272980 - 75289957 on Build GRCh38
- UniGene ID
- Hs.514470
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA743427.1 |
- |
1 |
145 |
|
BU607955.1 |
- |
2 |
154 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY228571.1 |
- |
1 |
177 |
|
Chr. 17 - 75272992 - 75273401 [+] |
GenBankmRNA |
BQ653688.1 |
- |
1 |
160 |
|
Chr. 17 - 75272992 - 75277479 [+] |
GenBankmRNA |
HY212150.1 |
- |
1 |
154 |
|
Chr. 17 - 75272992 - 75273406 [+] |
View Full Product Details