Gene Transcripts
- Gene Symbol
- LRPAP1
- Entrez Gene ID
- 4043
- Gene Name
- LDL receptor related protein associated protein 1
- Gene Aliases
- A2MRAP, A2RAP, HBP44, MRAP, MYP23, RAP, alpha-2-MRAP
- Chromosome Location
- Chr. 4: 3503597 - 3532497 on Build GRCh38
- UniGene ID
- Hs.40966
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC006574.1 |
- |
1 |
259 |
|
Chr. 4 - 3509185 - 3512409 [-] |
View Full Product Details