Gene Transcripts
- Gene Symbol
- Gm16006
- Entrez Gene ID
- 105245957
- Gene Name
- predicted gene 16006
- Gene Aliases
- OTTMUSG00000028937
- Chromosome Location
- Chr. 15: 57695426 - 57709946 on Build GRCm38
- UniGene ID
- Mm.438068
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042803.1 |
- |
2 |
142 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB640009.1 |
- |
2 |
142 |
|
Chr. 15 - 57708203 - 57708927 [-] |
View Full Product Details