Gene Transcripts
- Gene Symbol
- 1700012D14Rik
- Entrez Gene ID
- 75479
- Gene Name
- RIKEN cDNA 1700012D14 gene
- Gene Aliases
- Chromosome Location
- Chr. 7: 111117673 - 111122675 on Build GRCm38
- UniGene ID
- Mm.381620
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031468.1 |
- |
1 |
890 |
|
AK047116.1 |
- |
1 |
890 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV321778.2 |
- |
1 |
215 |
|
Chr. 7 - 111121322 - 111121995 [-] |
View Full Product Details