Gene Transcripts
- Gene Symbol
- Itgb1
- Entrez Gene ID
- 16412
- Gene Name
- integrin beta 1 (fibronectin receptor beta)
- Gene Aliases
- 4633401G24Rik, AA409975, AA960159, CD29, ENSMUSG00000051907, Fnrb, Gm9863, gpIIa
- Chromosome Location
- Chr. 8: 128685284 - 128733579 on Build GRCm38
- UniGene ID
- Mm.263396
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014611.1 |
- |
1 |
1163 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB236218.2 |
- |
1 |
332 |
|
Chr. 8 - 128684908 - 128685495 [-] |
GenBankmRNA |
BB684056.1 |
- |
1 |
257 |
|
Chr. 8 - 128685015 - 128685424 [-] |
View Full Product Details