Gene Transcripts
- Gene Symbol
- 2810427C15Rik
- Entrez Gene ID
- 69979
- Gene Name
- RIKEN cDNA 2810427C15 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013171.1 |
- |
1 |
342 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY712193.1 |
- |
1 |
348 |
|
Chr. 2 - 92306175 - 92307120 [+] |
View Full Product Details