Gene Transcripts

Gene Symbol
Herc4
Entrez Gene ID
67345
Gene Name
hect domain and RLD 4
Gene Aliases
1700056O17Rik, 4921531D01Rik, 9530080M15Rik, mKIAA1593
Chromosome Location
Chr. 10: 63243782 - 63317881 on Build GRCm38
UniGene ID
Mm.234437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020652.1 - 1 258


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CA325437.1 - 1 170 Chr. 10 - 63244236 - 63244924 [+]
GenBankmRNA BY722282.1 - 1 258 Chr. 10 - 63244148 - 63244557 [+]

View Full Product Details