Gene Transcripts
- Gene Symbol
- Gm16897
- Entrez Gene ID
- 320400
- Gene Name
- predicted gene, 16897
- Gene Aliases
- C030002c11rik
- Chromosome Location
- Chr. 1: 194955759 - 194976959 on Build GRCm38
- UniGene ID
- Mm.417608
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033564.1 |
|
5 |
1923 |
|
GenBank mRNA |
AK030377.1 |
- |
5 |
1925 |
|
View Full Product Details