Gene Transcripts
- Gene Symbol
- Myo5c
- Entrez Gene ID
- 208943
- Gene Name
- myosin VC
- Gene Aliases
- 9130003O20Rik, BB085560, C330029I24
- Chromosome Location
- Chr. 9: 75231725 - 75306263 on Build GRCm38
- UniGene ID
- Mm.41339
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK160451.1 |
- |
2 |
310 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707685.1 |
- |
2 |
306 |
|
Chr. 9 - 75232055 - 75233315 [+] |
View Full Product Details