Gene Transcripts
- Gene Symbol
- A330033J07Rik
- Entrez Gene ID
- 320614
- Gene Name
- RIKEN cDNA A330033J07 gene
- Gene Aliases
- Chromosome Location
- Chr. 13: 48044231 - 48262623 on Build GRCm38
- UniGene ID
- Mm.173009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102303.1 |
|
3 |
1971 |
|
GenBank mRNA |
AK034282.1 |
- |
3 |
1971 |
|
View Full Product Details