Gene Transcripts
- Gene Symbol
- 1700048B10Rik
- Entrez Gene ID
- 73465
- Gene Name
- RIKEN cDNA 1700048B10 gene
- Gene Aliases
- Chromosome Location
- Chr. 13: 50913817 - 50934152 on Build GRCm38
- UniGene ID
- Mm.418063
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006721.1 |
- |
1, 2 |
340 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB274787.1 |
- |
1, 2 |
107 |
|
Chr. 13 - 50914040 - 50933349 [-] |
View Full Product Details