Gene Transcripts
- Gene Symbol
- Mkln1os
- Entrez Gene ID
- 100503869
- Gene Name
- muskelin 1, intracellular mediator containing kelch motifs, opposite strand
- Gene Aliases
- Gm13845, OTTMUSG00000014733
- Chromosome Location
- Chr. 6: 31366885 - 31398773 on Build GRCm38
- UniGene ID
- Mm.370532
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK045552.1 |
- |
3 |
260 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB644004.1 |
- |
3 |
256 |
|
Chr. 6 - 31385878 - 31398757 [-] |
View Full Product Details