Gene Transcripts
- Gene Symbol
- Gm15759
- Entrez Gene ID
- 102636191
- Gene Name
- predicted gene 15759
- Gene Aliases
- OTTMUSG00000026037
- Chromosome Location
- Chr. 1: 58289410 - 58315656 on Build GRCm38
- UniGene ID
- Mm.411609
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028685.1 |
- |
|
3328 |
|
View Full Product Details