Gene Transcripts
- Gene Symbol
- Fam129a
- Entrez Gene ID
- 63913
- Gene Name
- family with sequence similarity 129, member A
- Gene Aliases
- AI256368, AU019833, Niban
- Chromosome Location
- Chr. 1: 151571373 - 151721939 on Build GRCm38
- UniGene ID
- Mm.157700
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK035217.1 |
- |
1 |
1047 |
|
AK080172.1 |
- |
1 |
543 |
|
View Full Product Details