Gene Transcripts
- Gene Symbol
- Gm1627
- Entrez Gene ID
- 381291
- Gene Name
- predicted gene 1627
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.328097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142343.1 |
- |
2 |
185 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY730468.1 |
- |
2 |
181 |
|
Chr. 1 - 133962165 - 133970003 [+] |
View Full Product Details