Gene Transcripts
- Gene Symbol
- 1700072F12Rik
- Entrez Gene ID
- 74278
- Gene Name
- RIKEN cDNA 1700072F12 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006936.1 |
- |
1 |
170 |
|
View Full Product Details