Gene Transcripts
- Gene Symbol
- 4930415P13Rik
- Entrez Gene ID
- 73948
- Gene Name
- RIKEN cDNA 4930415P13 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015156.1 |
- |
1 |
152 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY714703.1 |
- |
1 |
152 |
|
Chr. 18 - 46339896 - 46340429 [-] |
View Full Product Details