Gene Transcripts
- Gene Symbol
- 9330182O14Rik
- Entrez Gene ID
- 328531
- Gene Name
- RIKEN cDNA 9330182O14 gene
- Gene Aliases
- Chromosome Location
- Chr. 15: 40134365 - 40150010 on Build GRCm38
- UniGene ID
- Mm.60703
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034361.1 |
- |
4 |
1098 |
|
View Full Product Details