Gene Transcripts
- Gene Symbol
- B130055M24Rik
- Entrez Gene ID
- 654803
- Gene Name
- RIKEN cDNA B130055M24 gene
- Gene Aliases
- Chromosome Location
- Chr. 7: 127448654 - 127461238 on Build GRCm38
- UniGene ID
- Mm.423237
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041042.1 |
- |
2 |
1081 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB221467.2 |
- |
2 |
628 |
|
Chr. 7 - 127449089 - 127457185 [+] |
View Full Product Details