Gene Transcripts
- Gene Symbol
- Gm10714
- Entrez Gene ID
- 100038374
- Gene Name
- predicted gene 10714
- Gene Aliases
- ENSMUSG00000074555
- Chromosome Location
- Chr. 2: 174125038 - 174183918 on Build GRCm38
- UniGene ID
- Mm.127249
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040530.1 |
|
4 |
528 |
|
GenBank mRNA |
AK141020.1 |
- |
4 |
529 |
|
View Full Product Details