Gene Transcripts
- Gene Symbol
- Gm33100
- Entrez Gene ID
- 102635874
- Gene Name
- predicted gene, 33100
- Gene Aliases
- Chromosome Location
- Chr. 5: 96879310 - 96891170 on Build GRCm38
- UniGene ID
- Mm.244272
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK171414.1 |
- |
4 |
1160 |
|
View Full Product Details