Gene Transcripts
- Gene Symbol
- LOC101929596
- Entrez Gene ID
- 101929596
- Gene Name
- uncharacterized LOC101929596
- Gene Aliases
- Chromosome Location
- Chr. 2: 38458637 - 38515740 on Build GRCh38
- UniGene ID
- Hs.537341
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110259.1 |
|
5 |
1097 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000413227.1 |
- |
2 |
401 |
|
Chr. 2 - 38458646 - 38469441 [-] |
View Full Product Details