Gene Transcripts
- Gene Symbol
- Trpc5os
- Entrez Gene ID
- 100503240
- Gene Name
- transient receptor potential cation channel, subfamily C, member 5, opposite strand
- Gene Aliases
- ENSMUSG00000072934, Gm15070, OTTMUSG00000018931
- Chromosome Location
- Chr. X: 144455781 - 144477063 on Build GRCm38
- UniGene ID
- Mm.387258
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK147193.1 |
- |
4 |
2702 |
|
View Full Product Details