Gene Transcripts
- Gene Symbol
- Gm16892
- Entrez Gene ID
- 100503632
- Gene Name
- predicted gene, 16892
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.450368
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK149401.1 |
- |
4 |
1178 |
|
View Full Product Details