Gene Transcripts
- Gene Symbol
- 1700027F09Rik
- Entrez Gene ID
- 69999
- Gene Name
- RIKEN cDNA 1700027F09 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 64454813 - 64467942 on Build GRCm38
- UniGene ID
- Mm.261413
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040681.1 |
|
3 |
398 |
|
GenBank mRNA |
AK006413.1 |
- |
3 |
398 |
|
View Full Product Details