Gene Transcripts

Gene Symbol
LOC100506885
Entrez Gene ID
100506885
Gene Name
uncharacterized LOC100506885
Gene Aliases
Chromosome Location
Chr. 6: 19729421 - 19804759 on Build GRCh38
UniGene ID
Hs.346489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_134649.1 3 244
NR_134650.1 2 136
GenBank mRNA AA810378.1 - 2 126
AK092541.1 - 3 235

View Full Product Details