Gene Transcripts
- Gene Symbol
- LINC00700
- Entrez Gene ID
- 282980
- Gene Name
- long intergenic non-protein coding RNA 700
- Gene Aliases
- Chromosome Location
- Chr. 10: 2005473 - 2014348 on Build GRCh38
- UniGene ID
- Hs.576810
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040253.2 |
|
3 |
1911 |
|
GenBank mRNA |
AK097474.1 |
- |
3 |
1912 |
|
View Full Product Details