Gene Transcripts

Gene Symbol
TRAM2-AS1
Entrez Gene ID
401264
Gene Name
TRAM2 antisense RNA 1 (head to head)
Gene Aliases
Chromosome Location
Chr. 6: 52577202 - 52583993 on Build GRCh38
UniGene ID
Hs.544586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103446.1 1 1006
NR_103447.1 2 512
GenBank mRNA AK095117.1 - 1 901
BE741421.1 - 2 324

View Full Product Details