Gene Transcripts
- Gene Symbol
- CARMN
- Entrez Gene ID
- 728264
- Gene Name
- cardiac mesoderm enhancer-associated non-coding RNA
- Gene Aliases
- CARMEN, MIR143HG
- Chromosome Location
- Chr. 5: 149406845 - 149432836 on Build GRCh38
- UniGene ID
- Hs.519666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126541.1 |
- |
1 |
2950 |
|
BX640700.1 |
- |
2 |
1229 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF995194.1 |
- |
1 |
700 |
|
Chr. 5 - 149409520 - 149410269 [+] |
View Full Product Details