Gene Transcripts
- Gene Symbol
- MIR663AHG
- Entrez Gene ID
- 284801
- Gene Name
- MIR663A host gene
- Gene Aliases
- Chromosome Location
- Chr. 20: 26187019 - 26209233 on Build GRCh38
- UniGene ID
- Hs.370699
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040095.1 |
|
3 |
231 |
|
GenBank mRNA |
BC036544.1 |
- |
3 |
234 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000427348.5 |
- |
3 |
231 |
|
Chr. 20 - 26187019 - 26209233 [-] |
View Full Product Details