Gene Transcripts

Gene Symbol
Defb6
Entrez Gene ID
116746
Gene Name
defensin beta 6
Gene Aliases
BD-6
Chromosome Location
Chr. 8: 19225478 - 19228209 on Build GRCm38
UniGene ID
Mm.358822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054074.1 NP_473415.1 2 281
GenBank mRNA AB063109.1 - 2 281

View Full Product Details