Gene Transcripts

Gene Symbol
Dph1
Entrez Gene ID
116905
Gene Name
DPH1 homolog (S. cerevisiae)
Gene Aliases
2310011M22Rik, 4930488F09Rik, AW551873, Dph2l1, Ovca1
Chromosome Location
Chr. 11: 75177643 - 75190483 on Build GRCm38
UniGene ID
Mm.41496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144491.2 NP_652762.2 9 958
GenBank mRNA AK082812.1 - 9 963
AK128996.1 - 949
AY078170.1 - 9 949
BC022738.1 - 4 377
BC044663.1 - 9 929
BC080761.1 - 9 934

View Full Product Details