Gene Transcripts

Gene Symbol
Ube4a
Entrez Gene ID
140630
Gene Name
ubiquitination factor E4A
Gene Aliases
4732444G18Rik, 9930123J21Rik, UFD2b
Chromosome Location
Chr. 9: 44923127 - 44965600 on Build GRCm38
UniGene ID
Mm.215069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145400.3 NP_663375.3 16 2673
GenBank mRNA AK028721.1 - 16 2458
AK172893.1 - 16 2633
BC006649.1 - 18 2926 3498834

View Full Product Details