Gene Transcripts

Gene Symbol
Emid1
Entrez Gene ID
140703
Gene Name
EMI domain containing 1
Gene Aliases
AW122071, CO-5, Emu1
Chromosome Location
Chr. 11: 5106265 - 5152298 on Build GRCm38
UniGene ID
Mm.840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080595.2 NP_542162.1 3 240
GenBank mRNA AJ416093.1 - 3 240
AK153702.1 - 3 259
BC130023.1 - 3 245 40140754

View Full Product Details