Gene Transcripts

Gene Symbol
Plekhh2
Entrez Gene ID
213556
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 2
Gene Aliases
AI256725, E030001K05, mKIAA2028
Chromosome Location
Chr. 17: 84511895 - 84622142 on Build GRCm38
UniGene ID
Mm.215159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177606.4 NP_808274.2 25 3755
GenBank mRNA AK029252.1 - 25 3757
AK086794.1 - 5 585
AK173337.1 - 22 3410
BC125583.1 - 25 3720 40130098
BC137804.1 - 25 3717 8860823

View Full Product Details