Gene Transcripts

Gene Symbol
Scgn
Entrez Gene ID
214189
Gene Name
secretagogin, EF-hand calcium binding protein
Gene Aliases
Chromosome Location
Chr. 13: 23953456 - 23991337 on Build GRCm38
UniGene ID
Mm.255667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145399.1 NP_663374.1 10 859
GenBank mRNA AK148468.1 - 10 812
BC016093.1 - 10 859 4504330

View Full Product Details