Gene Transcripts

Gene Symbol
Impg2
Entrez Gene ID
224224
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, PG10.2, Rsbp, Spacrcan
Chromosome Location
Chr. 16: 56203842 - 56273753 on Build GRCm38
UniGene ID
Mm.151998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174876.3 NP_777365.2 15 3214
GenBank mRNA AK047356.1 - 1 105
AY174061.1 - 14 2887
BC026456.1 - 1 100
BC048863.1 - 15 3183 5368568

View Full Product Details