Gene Transcripts

Gene Symbol
Gtdc1
Entrez Gene ID
227835
Gene Name
glycosyltransferase-like domain containing 1
Gene Aliases
E330008O22Rik
Chromosome Location
Chr. 2: 44564412 - 44927918 on Build GRCm38
UniGene ID
Mm.314312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172662.3 NP_766250.2 6 997
GenBank mRNA AK028468.1 - 6 920
AK054272.1 - 6 998
BC052069.1 - 3 562 5715781

View Full Product Details