Gene Transcripts

Gene Symbol
Fbxl19
Entrez Gene ID
233902
Gene Name
F-box and leucine-rich repeat protein 19
Gene Aliases
6030430O11, BC059812, Fbl19
Chromosome Location
Chr. 7: 127744027 - 127769483 on Build GRCm38
UniGene ID
Mm.259353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172748.2 NP_766336.2 3 639
GenBank mRNA AK028405.1 - 3 408
AK031431.1 - 2 380
BC059812.1 - 3 639 6401846

View Full Product Details